Skip to Content
Merck
All Photos(1)

Key Documents

17-126

Sigma-Aldrich

PTP Assay Kit 2

Synonym(s):

protein tyrosine phosphatase assay kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

description

Phosphatase Assay

Quality Level

product line

Upstate®

Application

PTP Assay Kit 2 can be used to detect PTP-1B activity by either dephosphorylation of the phosphopeptide (T-S-T-E-P-Q-pY-Q-P-G-E-N-L) or hydrolysis of pNPP.

Quality

The components in this kit were used to detect PTP-1B (Catalog # 14-109) activity by either dephosphorylation of the phosphopeptide (T-S-T-E-P-Q-pY-Q-P-G-E-N-L) or hydrolysis of pNPP.

Other Notes

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3 - Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service