Key Documents
1.50170
Supelco
LiChroCART for customized packing HPLC Columns
L × I.D. 125 mm × 4 mm LiChroCART HPLC cartridge, manuCART holder required for use; please select HPLC sorbent for Column filling
About This Item
UNSPSC Code:
41104908
NACRES:
SB.52
product name
LiChroCART 125-4, HPLC cartridge
Quality Level
Legal Information
LICHROCART is a trademark of Merck KGaA, Darmstadt, Germany