1.17942
Phosphate Test Strips, reflectometric
0.1-5.0 mg/L (PO₄³⁻), for use with RQflex 10 Plus
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116116
NACRES:
NA.24
Recommended Products
Product Name
Phosphate Test, reflectometric, 0.1-5.0 mg/L (PO43-), for use with RQflex® 10 Plus
description
quantitative
Quality Level
specific analyte(s)
phosphate
measuring range
0.1-5.0 mg/L (PO43-)
compatibility
for use with RQflex® 10 Plus
detection method
reflectometric
storage temp.
15-25°C
General description
Method: reflectometric 0.1 - 5.0 mg/l PO Reflectoquant® plus or RQFlex® 10 plus
The Reflectoquant® Phosphate test measures orthophosphate in wastewater, drinking water, groundwater and surface water, aquarium water, seawater and food after appropriate sample pretreatment. In sulfuric solution orthophosphate ions (PO43-) react with molybdate ions to form molybdophosphoric acid. This is reduced to phosphomolybdenum blue (PMB) that is determined reflectometrically on the RQflex. For pre-testing a sample in an economic and quick way to estimate the phosphate concentration and a possible dilution factor prior to using this quantitative method we recommend our semi-quantitative MQuant test strips for phosphate. Further quantitative determination methods can be found with our Spectroquant tests.
Analysis Note
function controlconformsAssay
- standard 1
0.4 - 0.6 mg/l - standard 21.7 - 2.3 mg/l - standard 33.5 - 4.5 mg/l
- standard 1
0.4 - 0.6 mg/l - standard 21.7 - 2.3 mg/l - standard 33.5 - 4.5 mg/l
Legal Information
REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
RQFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A
Storage Class Code
8A - Combustible, corrosive hazardous materials
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service