Skip to Content
Merck
All Photos(1)

Key Documents

1.12576

Supelco

Chemizorb® Hg

reagents and accessories for absorption of mercury

Synonym(s):

Mercury Absorbent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131905
NACRES:
NB.51
Pricing and availability is not currently available.

Quality Level

form

powder

technique(s)

AAS: suitable

particle size

<63 μm

storage temp.

15-25°C

General description

Reagents and accessories for absorption of mercury

Analysis Note

Identity: passes test
Citric acid solution: about 20 %

Legal Information

CHEMIZORB is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service