Skip to Content
Merck
All Photos(2)

Key Documents

1.09309

Supelco

LiChrosorb® Si 100 (10 µm) HPLC Columns

HPLC sorbent, CAS 7631-86-9, EC Number 231-545-4, chemical formula SiO₂.

Synonym(s):

Silica gel

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41115709
EC Index Number:
231-545-4
NACRES:
SB.52
particle size:
10 μm
matrix active group:
Silica bonding phase
pore size:
100 Å pore size
matrix:
fully porous particle

Product Name

LiChrosorb® Si 100 (10 µm), LiChrosorb® Si 100 (10 µm). CAS 7631-86-9, EC Number 231-545-4, chemical formula SiO₂.

material

stainless steel column

Agency

suitable for USP L3

product line

LiChrosorb®

potency

>5000 mg/kg LD50, skin (Rabbit)

feature

endcapped: no

parameter

60 °C max. temp.

technique(s)

HPLC: suitable
mass spectrometry (MS): suitable (Low - Medium)

matrix

fully porous particle

matrix active group

Silica bonding phase

particle size

10 μm

pore size

100 Å pore size

operating pH

2-7.5 (20 °C, 100 g/L in H2O, slurry)

bp

2230 °C

mp

>1600 °C

bulk density

200‑800 kg/m3

separation technique

hydrophilic interaction (HILIC)
normal phase

storage temp.

no temp limit

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

LiChrosorb® Si 100 is a reliable irregular shaped silica sorbents with a pore diameter of 100A for normal phase chromatography.

Analysis Note

Pore volume: 0.98 - 1.06 ml/g
Specific surface area (BET): 290 - 340 m²/g
particle size (Malvern) (d10): 8.7 - 10.0 µm
particle size (Malvern) (d50): 12.0 - 14.0 µm
particle size (Malvern) (d90): 17.5 - 20.5 µm
Pressure: 10 - 16 bar
Theoretical Plates (N/m) (2-Nitrotoluene): ≥ 25000
Symmetry (2-Nitrotoluene): 0.8 - 1.4
Capacity factor (2-Nitrotoluene): 0.5 - 2.0

Legal Information

LiChrosorb is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service