1.07398
Lead(II) nitrate
for analysis EMSURE® ACS,Reag. Ph Eur
Synonym(s):
Lead(II) nitrate
About This Item
grade
ACS reagent
Quality Level
Agency
reag. Ph. Eur.
product line
EMSURE®
Assay
≥99.5% (complexometric)
form
solid
impurities
≤0.005% Insoluble matter
pH
3-4 (20 °C, 50 g/L in H2O)
mp
458-459 °C
solubility
486 g/L
density
4.49 g/cm3 at 20 °C
bulk density
1850 kg/m3
anion traces
chloride (Cl-): ≤0.0005%
cation traces
Ca: ≤0.005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Na: ≤0.02%
storage temp.
2-30°C
SMILES string
[Pb]1O[N+](=O)O1.[N+](=O)([O-])[O-]
InChI
1S/2NO3.Pb/c2*2-1(3)4;/q2*-1;+2
InChI key
RLJMLMKIBZAXJO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- Lead(II) nitrate and hexafluorosilicate complexes with neutral diphosphine coordination: The study investigates complexes of lead(II) nitrate with neutral diphosphine ligands, revealing detailed insights into their coordination chemistry and structural characteristics, which are crucial for developing new materials with enhanced performance (Burt J, Grantham W, Levason W, et al., 2015).
- Catalytically active lead(ii)-imidazolium coordination assemblies with diversified lead(ii) coordination geometries: This article describes the synthesis and characterization of lead(II) imidazolium coordination assemblies, which are noted for their diverse coordination geometries and catalytic activity, expanding the utility of lead(II) nitrate in catalytic applications (Naga Babu C, Suresh P, Srinivas K, et al., 2016).
- Removal of toxic heavy metal lead (II) using chitosan oligosaccharide-graft-maleic anhydride/polyvinyl alcohol/silk fibroin composite: This research focuses on the development of a bio-composite material for the effective removal of lead(II) from the environment, showcasing the application of chitosan oligosaccharide-graft-maleic anhydride/polyvinyl alcohol/silk fibroin composites in heavy metal remediation (P A, K V, M S, et al., 2017).
Linkage
Analysis Note
Insoluble matter: ≤ 0.005 %
Chloride (Cl): ≤ 0.0005 %
Ca (Calcium): ≤ 0.005 %
Cu (Copper): ≤ 0.0005 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.005 %
Na (Sodium): ≤ 0.02 %
corresponds to ACS,Reag. Ph Eur
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Repr. 1A - Skin Sens. 1B - STOT RE 1
Storage Class Code
5.1B - Oxidizing hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service