1.06591
tetra-Sodium diphosphate decahydrate
for analysis EMSURE® ACS,Reag. Ph Eur
Synonym(s):
tetra-Sodium diphosphate decahydrate, tetra-Sodium pyrophosphate, TSPP
About This Item
grade
ACS reagent
Quality Level
Agency
reag. Ph. Eur.
product line
EMSURE®
Assay
99.0-103.0% (acidimetric)
form
solid
impurities
≤0.001% Total nitrogen (N)
≤0.01% Insoluble matter
pH
9.5-10.5 (20 °C, 50 g/L in H2O)
mp
988 °C (anhydrous substance)
density
1.82 g/cm3 at 20 °C
bulk density
650 kg/m3
anion traces
chloride (Cl-): ≤0.001%
phosphate (PO43-): ≤0.1%
sulfate (SO42-): ≤0.005%
cation traces
Fe: ≤0.0005%
K: ≤0.005%
heavy metals (as Pb): ≤0.0005%
storage temp.
2-30°C
SMILES string
[Na+].[Na+].[Na+].[Na+].[P](=O)([O-])([O-])O[P](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O
InChI
1S/4Na.H4O7P2.10H2O/c;;;;1-8(2,3)7-9(4,5)6;;;;;;;;;;/h;;;;(H2,1,2,3)(H2,4,5,6);10*1H2/q4*+1;;;;;;;;;;;/p-4
InChI key
VZWGHDYJGOMEKT-UHFFFAOYSA-J
Looking for similar products? Visit Product Comparison Guide
Linkage
Analysis Note
Insoluble matter: ≤ 0.01 %
pH-value (5 %; water, 25 °C): 9.5 - 10.5
Chloride (Cl): ≤ 0.001 %
Orthophosphate (PO4): ≤ 0.1 %
Sulfate (SO₄): ≤ 0.005 %
Total nitrogen (N): ≤ 0.001 %
Heavy metals (as Pb): ≤ 0.0005 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.005 %
Corresponds to ACS, Reag. Ph Eur
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service