Skip to Content
Merck
All Photos(1)

Key Documents

1.06084

Supelco

Methylthymol blue sodium salt

metal indicator

Synonym(s):

Methylthymol blue sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H40N2Na4O13S
CAS Number:
Molecular Weight:
844.74
MDL number:
UNSPSC Code:
12171502
EC Index Number:
217-743-3
NACRES:
NA.05
Pricing and availability is not currently available.

Product Name

Methylthymol blue sodium salt, metal indicator

Quality Level

form

solid

loss

≤7% loss on drying, 50°C/ <1 hPa; 3h

pH

6.2 (20 °C, 10 g/L in H2O)

solubility

460 g/L

bulk density

570 kg/m3

Amax

430-450 nm (buffer pH 7.0)
602-606 nm (buffer pH 7.0)

storage temp.

2-30°C

SMILES string

[Na+].[Na+].[Na+].[Na+].[S]1(=O)(=O)OC(c4c1cccc4)(c3c(c(c(c(c3)C(C)C)O)CN(CC(=O)[O-])CC(=O)[O-])C)c2c(c(c(c(c2)C(C)C)O)CN(CC(=O)[O-])CC(=O)[O-])C

InChI

1S/C37H44N2O13S.4Na/c1-19(2)23-11-28(21(5)25(35(23)48)13-38(15-31(40)41)16-32(42)43)37(27-9-7-8-10-30(27)53(50,51)52-37)29-12-24(20(3)4)36(49)26(22(29)6)14-39(17-33(44)45)18-34(46)47;;;;/h7-12,19-20,48-49H,13-18H2,1-6H3,(H,40,41)(H,42,43)(H,44,45)(H,46,47);;;;/q;4*+1/p-4

InChI key

DIZZDZCUMBBRSG-UHFFFAOYSA-J

Analysis Note

Identity (UV/VIS-Spectrum): passes test
Absorption maximum λ 1 (buffer pH 7.0): 602 - 606 nm
Absorption maximum λ 2 (buffer pH 7.0): 430 - 450 nm
Spec. Absorptivity A 1%/1cm (λ1max; 0.04 g/l; buffer pH 7.0; calc. on dried substance): ≥ 60
Spec. Absorptivity A 1%/1cm (λ2max; 0.04 g/l; buffer pH 7.0; calc. on dried substance): ≥ 85
Loss on drying (50 °C/ <1 hPa; 3h): ≤ 7%
Suitability as indicator (for metal titration): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service