Skip to Content
Merck
All Photos(1)

Key Documents

1.04137

Sigma-Aldrich

Crystal violet (C.I. 42555)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H30ClN3
CAS Number:
Molecular Weight:
407.98
MDL number:
UNSPSC Code:
41116130
NACRES:
NA.21
Pricing and availability is not currently available.

form

solid

Quality Level

potency

420 mg/kg LD50, oral (Rat)

composition

Dye, ≥88% spectrophotometric assay

pH

2.5-3.5 (20 °C, 10 g/L in H2O)

mp

189-194 °C

solubility

10 g/L

density

1.19 g/L at 20 °C

bulk density

220‑400 kg/m3

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

InChI

1S/C25H30N3.ClH/c1-26(2)22-13-7-19(8-14-22)25(20-9-15-23(16-10-20)27(3)4)21-11-17-24(18-12-21)28(5)6;/h7-18H,1-6H3;1H/q+1;/p-1

InChI key

ZXJXZNDDNMQXFV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Crystal violet (C.I. 42555) for microscopy Certistain®, is a dry dye that can be used to produce a Crystal violet staining solution to differentiate Gram-positive and Gram-negative bacteria in samples of human origin. In the case of Gram-positive bacteria, the bacteria retain their dark blue / dark purple color even after the treatment with decolorizing solution. On the other side Gram-negative bacteria are stained red or pink with the counterstain. As CERTISTAIN® dye it is chemically quality-certified according to highest standards and strict specifications.

Application

for microscopy Certistain®

Analysis Note

Dye content (spectrophotometrically): ≥ 88%
Identity (UV/VIS-Spectrum): passes test
Absorption maximum λ max (water): 589 - 594 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.002 g/l; water): 2050 - 2450
TLC-Test: passes test
Michler`s ketone: <0.10%
Loss on drying (110 °C): ≤ 10%
Suitability for microscopy: passes test

Legal Information

CERTISTAIN is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service