Skip to Content
Merck
All Photos(2)

Key Documents

07-104

Sigma-Aldrich

Anti-IGF2 mRNA-binding protein 3 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

IGF II mRNA binding protein 3, IGF-II mRNA-binding protein 3, IGF2 mRNA-binding protein 3, KH domain containing protein overexpressed in cancer, KH domain-containing protein overexpressed in cancer, VICKZ family member 3, insulin-like growth factor 2 mRN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse, canine

species reactivity (predicted by homology)

rhesus monkey (based on 100% sequence homology), dog (based on 100% sequence homology), horse (based on 100% sequence homology), chimpanzee (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... IGF2BP3(10643)

General description

The protein encoded by this gene is primarily found in the nucleolus, where it can bind to the 5′ UTR of the insulin-like growth factor 2 leader 3 mRNA and may repress translation of insulin-like growth factor 2 during late development. The encoded protein contains several KH domains, which are important in RNA binding and are known to be involved in RNA synthesis and metabolism. A pseudogene exists on chromosome 7, and there are putative pseudogenes on other chromosomes. [provided by RefSeq].

Specificity

This antibody recognizes IGF2 mRNA-binding protein 3 at the C-terminus.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to human IGF2 mRNA-binding protein 3 at and around the C-terminus.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Growth Factors & Receptors
This Anti-IGF2 mRNA-binding protein 3 Antibody is validated for use in WB for the detection of IGF2 mRNA-binding protein 3.
Western Blot (SNAP ID) Analysis: 2 µg/mL from a previous lot detected IGF2 mRNA-binding protein 3 on 10 µg of Hek293 cell lysate.

Immunoprecipitation Analysis: A previous lot was used by an independent laboratory in IP.

Quality

Evaluated by Western Blot in Hek293 cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected IGF2 mRNA-binding protein 3 on 10 µg of Hek293 cell lysate.

Target description

64 kDa was observed. Predicted to cross-react with all the Isoforms of IGF2BP3.

Physical form

Affinity purified
Purified rabbit serum in PBS with 0.09% azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Hek293 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuhang Zhou et al.
Molecular cancer, 16(1), 77-77 (2017-04-13)
Gastric cancer (GC) is one of the frequent causes of cancer-related death in eastern Asian population. IGF2BP2 lists in the top rank up-regulated genes in GC, but its functional role is unclear. The expression of IGF2BP3 in GC cell lines
Insulin receptor substrate-2 regulates aerobic glycolysis in mouse mammary tumor cells via glucose transporter 1.
Shannon L Pankratz,Ernest Y Tan,Yumiko Fine,Arthur M Mercurio,Leslie M Shaw
The Journal of Biological Chemistry null
Dominik Schmiedel et al.
eLife, 5 (2016-03-18)
Expression of the stress-induced ligands MICA, MICB and ULBP 1-6 are up-regulated as a cellular response to DNA damage, excessive proliferation or viral infection; thereby, they enable recognition and annihilation by immune cells that express the powerful activating receptor NKG2D.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service