Skip to Content
Merck
All Photos(1)

Key Documents

V-056

Supelco

Riboflavin 5′-phosphate sodium salt solution

100 μg/mL (50:50 Methanol:1% Ammonium Acetate in Water, (as free phosphate)), certified reference material, ampule of 1 mL

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20N4O9P · xNa+
Molecular Weight:
455.34 (free acid basis)
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

product line

EMPLURA®

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

concentration

100 μg/mL (50:50 Methanol:1% Ammonium Acetate in Water, (as free phosphate))

format

single component solution

storage temp.

−70°C

Related Categories

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 3 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

75.2 °F

Flash Point(C)

24 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service