Skip to Content
Merck
All Photos(1)

Key Documents

V-035

Supelco

Nicotinic acid-13C6 solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Vitamin B3-13C6 solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6H5NO2
CAS Number:
Molecular Weight:
129.07
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing
clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

n1[13cH][13c]([13cH][13cH][13cH]1)[13C](=O)O

InChI

1S/C6H5NO2/c8-6(9)5-2-1-3-7-4-5/h1-4H,(H,8,9)/i1+1,2+1,3+1,4+1,5+1,6+1

InChI key

PVNIIMVLHYAWGP-IDEBNGHGSA-N

General description

Nicotinic acid, also known as niacin, is a water-soluble vitamin of the B complex and an essential human nutrient. Niacin deficiency, often the result of impaired nutritional status associated with conditions ranging from malnutrition to chronic alcoholism, is frequently monitored in patient plasma samples by LC-MS/MS.

Application

Nicotinic acid-13C6 is a stable-labeled internal standard suitable for quantification of nicotinic acid levels by MS-based testing applications from diagnostic testing of niacin deficiency to quality control testing of niacin-based pharmaceutical preparations, dietary supplements, or fortified foods.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service