Skip to Content
Merck
All Photos(1)

Key Documents

N-100

Supelco

Norchlordiazepoxide solution

1.0 mg/mL (Acetonitrile:DMSO (80:20)(v/v)), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12ClN3O
CAS Number:
Molecular Weight:
285.73
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL (Acetonitrile:DMSO (80:20)(v/v))

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

ClC1=CC2=C(N=C(N)C[N+]([O-])=C2C3=CC=CC=C3)C=C1

InChI

1S/C15H12ClN3O/c16-11-6-7-13-12(8-11)15(10-4-2-1-3-5-10)19(20)9-14(17)18-13/h1-8H,9H2,(H2,17,18)

InChI key

LNLMGKOHYYEWMY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Norchlordiazepoxide is a major urinary metabolite of the benzodiazepine chlordiazepoxide, indicated for short term treatment of severe anxiety and for the management of acute alcohol withdrawal syndrome. The metabolite, also known as N-desmethylchlordiazepoxide, is pharmacologically active. This Certified Spiking Solution® is suitable for GC/MS or LC/MS methods in chlordiazepoxide testing for forensic analysis, urine drug testing, or clinical toxicology.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyun-Sun Jeon et al.
Dental materials journal, 36(4), 476-481 (2017-04-20)
The aims of this study were to identify the optimal concentration of coated orthodontic elastomerics using a layer-by-layer technique that can release chlorhexidine (CHX) as an antimicrobial material, and to measure the physical properties and antimicrobial effects of the coated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service