Skip to Content
Merck
All Photos(1)

Key Documents

H-041

Supelco

(±)-11-Hydroxy-Δ9-THC-D3 solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27O3D3
CAS Number:
Molecular Weight:
333.48
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule D (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIB (Portugal)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

cannabis testing
forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

[2H]C([2H])([2H])CCCCc1cc(O)c2C3C=C(CO)CCC3C(C)(C)Oc2c1

InChI

1S/C21H30O3/c1-4-5-6-7-14-11-18(23)20-16-10-15(13-22)8-9-17(16)21(2,3)24-19(20)12-14/h10-12,16-17,22-23H,4-9,13H2,1-3H3/i1D3

InChI key

YCBKSSAWEUDACY-FIBGUPNXSA-N

General description

A stable-labeled internal standard suitable in LC/MS or GC/MS applications for cannabinoid testing, urine drug testing, clinical toxicology, or isotope dilution methods. 11-Hydroxy- Δ9-THC is an active metabolite of THC and metabolic precursor to 11-nor-9-carboxy-Δ9-THC. This cannabinoid biomarker is found in biological matrices including urine, blood, and plasma.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michal P Dybowski et al.
Forensic toxicology, 36(2), 415-423 (2018-07-03)
Analysis of drugs and their metabolites in biofluids usually demands the application of sample preparation methods that allow for full isolation of analyzed substances from the matrix. The purpose of this study was to develop a method using the QuEChERS

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service