Key Documents
Z285463
Sigma-Aldrich
PowrTrol heat controllers
AC/DC input 120 V AC, 10 A
About This Item
UNSPSC Code:
41112303
NACRES:
NB.76
Pricing and availability is not currently available.
manufacturer/tradename
Glas-Col 104APL120
availability
not available in EU
power consumption
1,200 W
General description
  • PowrTrol heat controller is a solid-state, proportional-voltage power control that provides precise manual control of mantles and tapes.
  • The output is adjustable from 3 to 100% of rated voltage.
  • The unit can be mounted to a lattice or used on bench top.
PowrTrol heat controllers includes:
Lighted On/Off switch, 10-amp output, Non-Linear control (scale is for reference only), Circuit protection, Low profile housing.
A solid-state, proportional-voltage power control that provides precise manual control of mantles and tapes. The output is adjustable from 3 to 100% of rated voltage. Can be mounted to a lattice or used on bench top.
Choose from one of the most recent versions:
Certificates of Analysis (COA)