Key Documents
Z282421
Sigma-Aldrich
Spinbar® assortment
polygon PTFE-coated, pack of 18 ea
About This Item
UNSPSC Code:
41103806
NACRES:
NB.72
Pricing and availability is not currently available.
material
polygon PTFE-coated
packaging
pack of 18 ea
manufacturer/tradename
Bel-Art F37184-0000
General description
2 of each size: 10 × 6 mm, 15 × 6 mm, 20 × 7 mm, 25 × 8 mm, 30 × 8 mm, 40 × 8 mm, 50 × 8 mm, 60 × 8 mm, 80 × 10 mm
Legal Information
Spinbar is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)