Key Documents
Z276235
Sigma-Aldrich
NMR pressure caps with long body
About This Item
UNSPSC Code:
41121800
NACRES:
SB.32
Pricing and availability is not currently available.
packaging
pk of 100 ea
color
red
General description
Style K-5.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. Hello, I want to know the raw material and size of this tube cap.
    1 answer
    1. The item is made of Low-density polyethylene. Please refer to the product drawing below. The dimensions by letter designation are as follows:
      A = 0.227in.
      B = 0.272in.
      C = 0.31in.
      D = 0.14in.
      E = 0.38in.
      F:0.20in.
      Helpful?