Key Documents
Z257400
Spirit-filled low-temperature thermometer
L 305 mm (30.5 cm), temp. range -100/50 °C
About This Item
UNSPSC Code:
41112303
NACRES:
NB.79
Pricing and availability is not currently available.
packaging
pack of 1 ea
manufacturer/tradename
HB Instruments B60760-1400
parameter
-100/50 °C temp. range
L
305 mm (30.5 cm)
General description
These thermometers contain an organic liquid except Z257419 which is filled with a clear Toluene liquid. Total-immersion. 1° divisions for use in general-laboratory, low-temperature applications.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. the description says the thermometer is filled with organic liquid, but doesn't specify what liquid that is. Do you have that information or a safety sheet for the liquid? I'd just like to know how toxic the substance is, e.g. if it contains toluene
    1 answer
    1. This item is a product of Bel-Art. The thermometer contains toluene, as indicated on the manufacturer's Safety Data Sheet (SDS). Please see the link below to access the SDS:
      https://www.belart.com/media/catalogstudio/SDS/Toluene.pdf
      Helpful?