Skip to Content
Merck
All Photos(1)

Documents

W412901

Sigma-Aldrich

1-Hepten-3-ol

≥98%, FG

Synonym(s):

Butyl vinyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH(OH)CH=CH2
CAS Number:
Molecular Weight:
114.19
FEMA Number:
4129
Beilstein:
1720340
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.155
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 872/2012

Assay

≥98%

refractive index

n20/D 1.433 (lit.)

bp

153-156 °C (lit.)

density

0.836 g/mL at 20 °C
0.836 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; mushroom; fruity; earthy

SMILES string

CCCCC(O)C=C

InChI

1S/C7H14O/c1-3-5-6-7(8)4-2/h4,7-8H,2-3,5-6H2,1H3

InChI key

PZKFYTOLVRCMOA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

128.8 °F - closed cup

Flash Point(C)

53.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sándor Kecskeméti et al.
Journal of chemical ecology, 46(10), 917-926 (2020-10-08)
The chemical signatures emitted by fungal substrates are key components for mycophagous insects in the search for food source or for suitable oviposition sites. These volatiles are usually emitted by the fruiting bodies and mycelia. The volatiles attract fungivorous insects
Pingxi Xu et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(46), 16592-16597 (2014-10-29)
Insect repellents are important prophylactic tools for travelers and populations living in endemic areas of malaria, dengue, encephalitis, and other vector-borne diseases. DEET (N,N-diethyl-3-methylbenzamide) is a 6-decade-old synthetic repellent, which is still considered the gold standard of mosquito repellents. Mosquitoes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service