Key Documents
SYND381412
Synthware distilling receiver with 12 mL receiving tubes
joint: ST/NS 14/20
Synonym(s):
Synthware D381412, multi-tube distillation receiver
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Pricing and availability is not currently available.
Joint
joint: ST/NS14/20
packaging
pack of 1 ea
manufacturer/tradename
D381412
Related Categories
General description
Synthware distillation receiver features a standard taper joint (14/20) and three 12 mL receiving tubes graduated connected by three plastic joint clips with a 135° angle between the top outer joint and the lower inner joint.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)