SYNA791924
Synthware™ bushing adapter
female joint: ST/NS 19/38, male joint: ST/NS 24/40
Synonym(s):
Synthware A791924, glass bushing adapter
All Photos(1)
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Recommended Products
Joint
female joint: ST/NS19/38
male joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A791924
General description
Bushing adapters allow for compact assemblies of dissimilar joints.
Legal Information
Synthware is a trademark of Kemtech America Inc.