Skip to Content
Merck
All Photos(1)

Key Documents

QBD10224

Sigma-Aldrich

Bis-dPEG®5-NHS ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H32N2O13
CAS Number:
Molecular Weight:
532.50
MDL number:
UNSPSC Code:
51171641
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

>90%

form

solid or viscous liquid

reaction suitability

reagent type: cross-linking reagent
reactivity: amine reactive

polymer architecture

shape: linear
functionality: homobifunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(CCOCCOCCOCCOCCOCCC(ON1C(CCC1=O)=O)=O)ON2C(CCC2=O)=O

InChI

1S/C22H32N2O13/c25-17-1-2-18(26)23(17)36-21(29)5-7-31-9-11-33-13-15-35-16-14-34-12-10-32-8-6-22(30)37-24-19(27)3-4-20(24)28/h1-16H2

InChI key

FTYUGLBWKRXQBD-UHFFFAOYSA-N

Features and Benefits

Bis-dPEG®5-NHS ester is a homobifunctional dPEG® crosslinking product which allows conjugation to two different moieties, each containing free amines. The single molecular weight dPEG® spacer provides precise distance control between conjugated moieties. N-hydroxysuccinimide (NHS) ester versions are highly reactive to free amines.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service