Skip to Content
Merck
All Photos(1)

Key Documents

L040080

Sigma-Aldrich

Cap A

configured for PerkinElmer, configured for Polygen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.21
Pricing and availability is not currently available.

type

for DNA synthesis

Quality Level

product line

Proligo Reagents

form

liquid

storage condition

dry at room temperature

concentration

88.7-108.4 g/L (Acetic Anhydride content, titration)

color

clear

compatibility

configured for PerkinElmer
configured for Polygen

Looking for similar products? Visit Product Comparison Guide

General description

Cap A solution contains acetic anhydride and tetrahydrofuran (THF). It is used after the phosphoramidite reaction/coupling step.

Biochem/physiol Actions

Cap A solution aborts chains bearing unreacted 5′ hydroxyl groups due to failure of reaction with activated phosphoramidite (typically, 1 to 2% of growing oligonucleotide chains in each synthesis cycle). These unreacted 5′ hydroxyl groups are capped with an acetyl group and thus rendered unreactive for subsequent synthesis steps.

Preparation Note

Acetic Anhydride / Tetrahydrofurane 9.1/90.9 (v/v)

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-6.2 °F - closed cup

Flash Point(C)

-21.2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service