Key Documents
HAM201050
Hamilton® Gastight® syringe, general use, 1700 series
model, 1710, AccuDil (AD) end, volume 100 μL
About This Item
UNSPSC Code:
41122001
NACRES:
NB.03
Pricing and availability is not currently available.
material
Borosilicate glass/PTFE
Gastight syringe
feature
autoclavable: no
model 1710, AccuDil (AD) end
manufacturer/tradename
Hamilton
parameter
1000 psig pressure
20-50 °C temp. range (68-122 °F)
volume
100 μL
Legal Information
GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)