Skip to Content
Merck
All Photos(2)

Key Documents

GF65118630

Zirconium

foil, 6mm disks, thickness 0.01mm, 99.2%

Synonym(s):

Zirconium, ZR000202

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Zr
CAS Number:
Molecular Weight:
91.22
MDL number:
UNSPSC Code:
12141751
PubChem Substance ID:
NACRES:
NA.23

Assay

99.2%

form

foil

manufacturer/tradename

Goodfellow 651-186-30

resistivity

40 μΩ-cm, 20°C

diam. × thickness

6 mm × 0.01 mm

bp

4377 °C (lit.)

mp

1852 °C (lit.)

density

6.5 g/mL at 25 °C (lit.)

SMILES string

[Zr]

InChI

1S/Zr

InChI key

QCWXUUIWCKQGHC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abnormal trace metals in man: zirconium.
H A Schroeder et al.
Journal of chronic diseases, 19(5), 573-586 (1966-05-01)
Oluwatayo F Ikotun et al.
Future medicinal chemistry, 3(5), 599-621 (2011-04-30)
Positron emission tomography, with its high sensitivity and resolution, is growing rapidly as an imaging technology for the diagnosis of many disease states. The success of this modality is reliant on instrumentation and the development of effective and novel targeted
Yugo Hatai
The international journal of esthetic dentistry, 9(2), 206-222 (2014-04-26)
In contemporary dentistry, we have a vast range of materials to choose from, and metal free restorations have become the premier materials for achieving the ultimate in both esthetics and durability. Metal-free restorations are utilized with more conservative preparations to
Glypican-3-targeted 89Zr PET imaging of hepatocellular carcinoma: where antibody imaging dares to tread.
Bernadette V Marquez et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 55(5), 708-709 (2014-03-26)
Anjana Raut et al.
Indian journal of dental research : official publication of Indian Society for Dental Research, 22(1), 140-143 (2011-04-29)
The demand for esthetic restorations has resulted in an increased use of dental ceramics for anterior and posterior restorations. A few decades ago, all-ceramic restorations were restricted to treatment in the anterior region, but now all-ceramic restorations can be made

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service