Skip to Content
Merck
All Photos(1)

Key Documents

GF47476637

Titanium carbide

powder, mean particle size (fsss) 3 micron, weight 50 g, purity 98.5%

Synonym(s):

TI546010

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352103
NACRES:
NA.23

Assay

98.5%

form

powder

manufacturer/tradename

Goodfellow 474-766-37

weight

50 g

mean particle size

3 μm (FSSR)

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service