GF42273266
Stainless steel - AISI 304L alloy, Fe/Cr18/Ni10
foil, 100 x 200mm, 0.9mm thickness, annealed, mirror polished on one side
Synonym(s):
FA140501
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
11171600
NACRES:
NA.23
form
annealed (mirror polished on one side)
foil
manufacturer/tradename
Goodfellow 422-732-66
size × thickness
100 mm × 200 mm × 0.9 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service