There is no indication that this product has been polished. Polystyrene sheets typically do not require polishing for general use, as they usually come with a smooth surface finish suitable for many applications.
GF23575686
Polystyrene
(PS), sheet, thickness 2 mm, size 300 × 300 mm, condition crystal
Synonym(s):
ST313200
About This Item
Recommended Products
form
sheet
manufacturer/tradename
Goodfellow 235-756-86
size
300 × 300 mm
thickness
2 mm
General description
Legal Information
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Is the GF23575686 polystyrene sheet polished?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service