GF20050108
Gold-palladium alloy, Au93Pd7
0.1m wire, 0.5mm diameter, as drawn
Synonym(s):
AC015140
Select a Size
Select a Size
About This Item
Recommended Products
form
wire
manufacturer/tradename
Goodfellow 200-501-08
L × diam.
0.1 m × 0.5 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service