Skip to Content
Merck
All Photos(1)

Key Documents

GF16609057

Alumina

foam, thickness 7 mm, size 50 × 50 mm, porosity 0.86

Synonym(s):

AL603820

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11181511
NACRES:
NA.23

Pricing and availability is not currently available.

manufacturer/tradename

Goodfellow 166-090-57

size

50 × 50 mm

thickness

7 mm

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF00710750GF95382771GF44242118
size

50 × 50 mm

size

30 × 100 mm

size

-

size

100 × 100 mm

thickness

7 mm

thickness

7 mm

thickness

12.7 mm

thickness

7 mm

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Sorry, we don't have COAs for this product available online at this time.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service