Key Documents
DWK90P20113F
KIMBLE® disposable screw thread culture tube
flat bottom borosilicate glass, tube diam. × L (20 mm) (113 mm), non-sterile
Synonym(s):
90P20113F
About This Item
UNSPSC Code:
41121706
NACRES:
NB.11
material
flat bottom borosilicate glass
flat bottom
screw top neck
transparent borosilicate glass tube
sterility
non-sterile
feature
without cap
packaging
case of 500 ea Modular tray shrink-wrapped in plastic film
manufacturer/tradename
Kimble® 90P20113F
tube diam. × L
(20 mm) (113 mm)
fitting
neck (External Thread)
General description
Disposable Screw Thread Culture Tubes
  • These tubes are suitable for tissue culture work and general bacteriological use.
  • Modular trays are shrink-wrapped in plastic film
  • Supplied without caps
  • No marking spot
  • Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
Features and Benefits
  • Manufactured from borosilicate glass 5.1
  • 73760 series and 90 series items that end in ‘F′ have flat bottoms which contribute to stability
  • 73770 series items are produced with a round bottom
  • Modular trays are shrink-wrapped in plastic film
  • Supplied without caps
  • No marking spot
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)