Skip to Content
Merck
All Photos(1)

Documents

D55407

Sigma-Aldrich

2,3-Dichloroaniline

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3NH2
CAS Number:
Molecular Weight:
162.02
Beilstein:
472027
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

refractive index

n20/D 1.597 (lit.)

bp

252 °C (lit.)

mp

20-25 °C (lit.)

density

1.37 g/mL at 20 °C (lit.)

SMILES string

Nc1cccc(Cl)c1Cl

InChI

1S/C6H5Cl2N/c7-4-2-1-3-5(9)6(4)8/h1-3H,9H2

InChI key

BRPSAOUFIJSKOT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

239.0 °F - closed cup

Flash Point(C)

115 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service