Skip to Content
Merck
All Photos(1)

Key Documents

D145807

Sigma-Aldrich

2,3-Dimethylaniline

99%

Synonym(s):

1-Amino-2,3-dimethylbenzene, 2,3-Xylidine, 3-Amino-o-xylene, CN-Cbl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3NH2
CAS Number:
Molecular Weight:
121.18
Beilstein:
742174
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

4.2 (vs air)

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.569 (lit.)

bp

221-222 °C (lit.)

mp

2.5 °C (lit.)

density

0.993 g/mL at 25 °C (lit.)

SMILES string

Cc1cccc(N)c1C

InChI

1S/C8H11N/c1-6-4-3-5-8(9)7(6)2/h3-5H,9H2,1-2H3

InChI key

VVAKEQGKZNKUSU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dilek Arpaci et al.
Journal of ovarian research, 8, 71-71 (2015-11-08)
Polycystic ovary syndrome (PCOS) is related to metabolic syndrome, insulin resistance, and cardiovascular metabolic syndromes. This is particularly true for individuals with central and abdominal obesity because visceral abdominal adipose tissue (VAAT) and epicardial adipose tissue (EAT) produce a large
Stéphanie Romand et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 63, 14-21 (2014-07-06)
A capillary isoelectric focusing (cIEF) method was developed for the determination of the ionization constants (pKa) of small molecules. Two approaches used to decrease the electroosmotic flow (EOF) were compared: (i) a hydroxypropylcellulose (HPC) coated capillary in aqueous medium and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service