Skip to Content
Merck
All Photos(1)

Key Documents

CLS76309

Pyrex® PTFE key-hole stopper, color-coded

black, stopper size 9

Synonym(s):

flask stopper, flask stoppers, stoppers, Corning® 7630

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122500
NACRES:
NB.43

stopper size

9

manufacturer/tradename

Corning 7630-9

color

black

Looking for similar products? Visit Product Comparison Guide

General description

These chemically resistant Pyrex keyhole stoppers are made are made from PTFE and conform to stopper dimensions. The color-coded keys make sorting and selecting the correct size much faster and easier. These stoppers may be used in place of conventional glass flask stoppers

Legal Information

Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service