CLS7078B25
Pyrex® Shorty serological disposable pipette, sterile, plugged
volume 25.0 mL, accuracy: 0.2 mL
Synonym(s):
Corning® 7078B
About This Item
Recommended Products
material
glass
round tip
sterility
sterile
feature
color-coded Red
manufacturer/tradename
Corning 7078B-25
volume measuring range
25.0 mL, accuracy: 0.2 mL
L
300 mm
capacity
25 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service