C88602
Cyanoacetohydrazide
98%
Synonym(s):
Cyanoacetic hydrazide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
NCCH2CONHNH2
CAS Number:
Molecular Weight:
99.09
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
powder
mp
108-110 °C (lit.)
SMILES string
NNC(=O)CC#N
InChI
1S/C3H5N3O/c4-2-1-3(7)6-5/h1,5H2,(H,6,7)
InChI key
HPHBOJANXDKUQD-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
X M He et al.
Acta crystallographica. Section A, Foundations of crystallography, 49 ( Pt 1), 10-22 (1993-01-01)
For molecular crystals, a procedure is proposed for interpreting experimentally determined atomic mean square anisotropic displacement parameters (ADPs) in terms of the overall molecular vibration together with internal vibrations with the assumption that the molecule consists of a set of
S H Doss et al.
Archives of pharmacal research, 24(5), 377-384 (2001-11-06)
The reaction of either Digitoxin or Digoxin with cyanoacetic acid hydrazide gave the hydrazone derivatives 3a and 3b respectively. The reactivity of the latter products towards chemical reagents was studied to give heterocyclic derivatives with potential biological activities.
Soad A M El-Hawash et al.
Archiv der Pharmazie, 339(1), 14-23 (2006-01-18)
Two new acetylpyridinehydrazones derived from cyanoacetic acid hydrazide have been synthesized namely: cyanoacetic acid (1-pyridin-3 or 4-yl-ethylidene) hydrazides (1a,b). and some derived ring systems: 2-imino or 2-oxo-2H-chromenes (2a,b and 3a,b), substituted 2-thioxo-2,3-dihydrothiazoles (4a-d), substituted 2-thioxo-2,3-dihydro-6H-thiazolo[4,5-d]pyrimidin-7-ones (5a-d), substituted dihydrothiazoles (7a,b), and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service