Key Documents
BR708745
BRAND® disposable BLAUBRAND® micropipettes, intraMark
tube capacity 50-100 μL , Dual marks, blue
Synonym(s):
Micropipettes, Pipettes, intraMARK
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
Pricing and availability is not currently available.
material
blue
feature
DE-M marking, To Contain
packaging
pack of 1000 ea (4 boxes of 250)
manufacturer/tradename
BRAND 708745
L
125 mm
tube capacity
50-100 μL , Dual marks
General description
DIN ISO 7550. DE-M marking for volumes greater than 5 µl. Calibrated to contain (TC, In). Color code to ISO for clear identification.
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)