BR701365
BRAND® sealing film for microplates
self-adhesive, gas permeable, suitable for (for cell and tissue culture)
Synonym(s):
Accessories, PCR, Plates, Sealing films
All Photos(1)
About This Item
UNSPSC Code:
41122410
NACRES:
NB.22
Recommended Products
material
Rayon
Rayon film
sterility
sterile
feature
gas permeable
self-adhesive
packaging
pack of 50 ea
manufacturer/tradename
BRAND 701365
suitability
suitable for (for cell and tissue culture)
Related Categories
General description
When working with cell and tissue cultures, ensuring a secure seal is only the first step in protecting against contamination and facilitating safe transport. Equally critical is the ability to allow for proper gas exchange. Insufficient oxygen levels or excessive carbon dioxide can not only compromise the accuracy of experimental results but may also jeopardize cell viability. Consequently, films designed for cell and tissue cultures must meet stringent requirements to support both secure sealing and optimal gas exchange, ensuring a conducive environment for cellular growth and experimentation.
Features and Benefits
- Air permeable for optimum oxygen supply
- Secure seal against contamination
- Pierceability for easy sample extraction
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)