Key Documents
BR61750
BRAND® sterilization indicator tape
crepe paper, self-adhesive
Synonym(s):
Sterilization indicator tape
About This Item
UNSPSC Code:
55121611
NACRES:
NB.35
Pricing and availability is not currently available.
material
paper
feature
crepe paper
self-adhesive
packaging
pkg of 1 roll
manufacturer/tradename
BRAND 61750
size
50 m × 19 mm
General description
Containing thermo-active pigments. The white colored strips turn brown after autoclaving (121°C: after 20 min. approx., 134°C: after 5 min. approx.).
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Related Content
Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.
Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.
Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.
Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.