Key Documents
BR134002
BRAND® non-return valve
for tube I.D. 6-9 mm
Synonym(s):
Non-return valve, Tubing
About This Item
UNSPSC Code:
40141600
NACRES:
NB.35
material
high-density polyethylene
packaging
pack of 10 ea
manufacturer/tradename
BRAND 134002
L
80 mm
Related Categories
General description
For tubing of inner- 6-9 mm. Length 80 mm. With FKM valve disc not suitable for pressure applications.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)