Skip to Content
Merck
All Photos(3)

Key Documents

A902

Sigma-Aldrich

Acetaldehyde diethyl acetal

99%

Synonym(s):

1,1-Diethoxyethane, Acetal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OC2H5)2
CAS Number:
Molecular Weight:
118.17
Beilstein:
1098310
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

4.1 (vs air)

Quality Level

vapor pressure

20 mmHg ( 20 °C)
8 mmHg ( 8 °C)

Assay

99%

form

liquid

autoignition temp.

446 °F

expl. lim.

10.4 %

refractive index

n20/D 1.379-1.383 (lit.)

bp

102 °C (lit.)

density

0.831 g/mL at 25 °C (lit.)

SMILES string

CCOC(C)OCC

InChI

1S/C6H14O2/c1-4-7-6(3)8-5-2/h6H,4-5H2,1-3H3

InChI key

DHKHKXVYLBGOIT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Acetaldehyde diethyl acetal can react with diketene to form ethyl 5-ethoxy-3-oxohexanoate in the presence of titanium chloride.[1] It can also be used to synthesize mixed acetal glycosides via transacetalation.[2]

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

<68.0 °F - closed cup

Flash Point(C)

< 20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A convenient method for the synthesis of δ-alkoxy-β-ketoester the titanium tetrachloride-activated reaction of diketene with acetal.
Izawa T and Mukaiyama T
Chemistry Letters (Jpn), 3(10), 1189-1192 (1974)
Glycosides for testing glycosidases: Vinyl, l-ethoxyethyl, and l-ethoxybut-3-enyl d-glucopyranosides.
Dettinger H M, et al.
Carbohydrate Research, 87(1), 63-70 (1980)
Geelsu Hwang et al.
Biofouling, 30(9), 1079-1091 (2014-10-31)
Well-established biofilms formed by Streptococcus mutans via exopolysaccharide matrix synthesis are firmly attached to tooth surfaces. Enhanced understanding of the physical properties of mature biofilms may lead to improved approaches to detaching or disassembling these highly organized and adhesive structures.
A J Cappadoro et al.
Animal reproduction science, 158, 104-108 (2015-05-30)
The purpose of this study was to develop a new injection-molded intravaginal insert manufactured from ethylene-vinyl acetate containing progesterone for a 7-day insertion period in cattle. The manufacturing process resulted in a reduction in the residual drug compared to the
Federica Sangiuolo et al.
The European respiratory journal, 45(2), 483-490 (2014-12-17)
In idiopathic pulmonary fibrosis (IPF), lung accumulation of excessive extracellular iron and macrophage haemosiderin may suggest disordered iron homeostasis leading to recurring microscopic injury and fibrosing damage. The current study population comprised 89 consistent IPF patients and 107 controls. 54

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. S'il vous plaît le mécanisme de synthèse comment le fais t'on

    1 answer
    1. This product has a quality level 200. The methods or preparation is available for products with quality 400 and above. The synthesis mechanism is considered a method of preparation which is proprietary.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service