Skip to Content
Merck
All Photos(1)

Key Documents

936391

Sigma-Aldrich

Fmoc-Gly-Gly-OH

≥95%

Synonym(s):

Fmoc-Gly-Gly-OH, 2-[2-([[(9H-Fluoren-9-yl)methoxy]carbonyl]amino)acetamido]acetic acid, N-9-Fluorenylmethoxycarbonylglycylglycine, N-Fluorenylmethoxycarbonylglycylglycine, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]glycylglycine (ACI), Glycine, N-[(9H-fluoren-9-ylmethoxy)carbonyl]glycyl- (9CI, ACI), Glycine, N-[N-[(9H-fluoren-9-ylmethoxy)carbonyl]glycyl]- (ZCI)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18N2O5
CAS Number:
Molecular Weight:
354.36
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder or crystals

color

white to off-white

storage temp.

2-8°C

SMILES string

O=C(OCC1C=2C=CC=CC2C=3C=CC=CC31)NCC(=O)NCC(=O)O

InChI

1S/C19H18N2O5/c22-17(20-10-18(23)24)9-21-19(25)26-11-16-14-7-3-1-5-12(14)13-6-2-4-8-15(13)16/h1-8,16H,9-11H2,(H,20,22)(H,21,25)(H,23,24)

Application

Fmoc-Gly-Gly-OH is a lysosomally cleavable linker used in construction of antibody-drug conjugates (ADC & Bioconjugation).

Other Notes

Chemical Biology Tools

http://pubs.rsc.org/en/Content/ArticleLanding/2011/MB/C0MB00074D#!divAbstract

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alain Beck et al.
Nature reviews. Drug discovery, 16(5), 315-337 (2017-03-18)
Antibody-drug conjugates (ADCs) are one of the fastest growing classes of oncology therapeutics. After half a century of research, the approvals of brentuximab vedotin (in 2011) and trastuzumab emtansine (in 2013) have paved the way for ongoing clinical trials that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service