Skip to Content
Merck
All Photos(1)

Key Documents

929506

Sigma-Aldrich

ItOct-bimy HCl

≥95%

Synonym(s):

1,3-Bis(2,4,4-trimethylpentan-2-yl)-1H-benzo[d]imidazol-3-ium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H39ClN2
Molecular Weight:
379.02
MDL number:
UNSPSC Code:
12352005
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder

SMILES string

CC(C)(CC(C)(C)C)N1C=[N+](C(C)(CC(C)(C)C)C)C2=C1C=CC=C2.[Cl-]

InChI

1S/C23H39N2.ClH/c1-20(2,3)15-22(7,8)24-17-25(19-14-12-11-13-18(19)24)23(9,10)16-21(4,5)6;/h11-14,17H,15-16H2,1-10H3;1H/q+1;/p-1

Application

ItOct-bimy HCl is an N-alkyl NHC ligand with demonstrated uses in the following reactions. Au-catalyzed hydration, Cu-catalyzed hydroboration, Ag-catalyzed hydroboration, Cu-catalyzed C–O coupling and Pd-catalyzed C–C and C–N couplings. This ligand features significantly higher steric volume compared with ItBu-bimy (673390) while maintaining the σ-donation and σ-acceptance inherent to N-alkyl NHC ligands.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service