919225
CdSe/ZnS core-shell type quantum dots
oleic acid functionalized, fluorescence λem 620 nm, 5 mg/mL in toluene
Synonym(s):
Fluorescent nanocrystals, QDs
About This Item
description
stabilized with oleic acid
Quality Level
form
liquid
concentration
5 mg/mL in toluene
fluorescence
FWHM 23-28 nm
λem 620 nm
, quantum yield ≥70
functional group
oleic acid
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
Application
Caution
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3
Target Organs
Central nervous system
WGK
WGK 3
Flash Point(F)
39.2 °F
Flash Point(C)
4 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service