Skip to Content
Merck
All Photos(1)

Key Documents

906344

Sigma-Aldrich

PBDB-T-SF

Mw ≥80,000

Synonym(s):

OS0130, PDBD-TSF, Poly[(2,6-(4,8-bis(5-(2-ethylhexylthio)-4-fluorothiophen-2-yl)-benzo[1,2-b:4,5-b′]dithiophene))-alt-(5,5-(1′,3′-di-2-thienyl-5′,7′-bis(2-ethylhexyl)benzo[1′,2′-c:4′,5′-c′]dithiophene-4,8-dione)]

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C68H76F2O2S10)n
UNSPSC Code:
12352101
NACRES:
NA.23

description

Band gap: 1.8 eV

form

granular

mol wt

Mw ≥80,000

color

purple, Shine

solubility

chlorobenzene: soluble
chloroform: soluble
soluble (o-dichlorobenzene)

Orbital energy

HOMO -5.4 eV 
LUMO -3.6 eV 

PDI

~2.5

Application

PBDB-T-SF is a fluorinated wide-bandgap polymeric donor (n-type semiconductor) with relatively deep highest occupied molecular orbital (HOMO) energy level. It has been used in high performance polymer organic solar cells (PSCs).
HOMO =−5.40eV
LUMO =−3.60eV

Polymer organic solar cells (PSCs) device performance:

Device based on PBDB-T-SF:NCBDT-4Cl
Before any post-treatment
PCE= of 13.1%
After device optimizations
Voc = 0.85V
Jsc = 2.35 mA /cm2
FF = 74.3%
PCE >14%
Energy loss = 0.55 eV

The improved performance was attributed to the more efficient photo-electron conversion process in the optimal device.

Device performance based on PBDB-T-SF:ITIC-F as active layer
(ITO)/ZnO/active layer/MoO3/Al
Voc = 0.88 V
Jsc = 20.50 mA/cm2
FF = 71.9%
PCE = 13.1%
Energy loss = 0.66 eV

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Molecular Optimization Enables over 13% Efficiency in Organic Solar Cells
Z Wenchao, et al.
Journal of the American Chemical Society, 139(21), 7148-7151 (2017)
A chlorinated low-bandgap small-molecule acceptor for organic solar cells with 14.1% efficiency and low energy loss.
Kan B, et al.
Science China: Chemistry, 61(10), 1307-1313 (2018)

Articles

Professor Chen (Nankai University, China) and his team explain the strategies behind their recent record-breaking organic solar cells, reaching a power conversion efficiency of 17.3%.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service