Skip to Content
Merck
All Photos(1)

Key Documents

903655

Sigma-Aldrich

1-Cyclohexyl-2-norbornene

Synonym(s):

1-Cyclohexylbicyclo[2.2.1]hept-2-ene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H20
CAS Number:
Molecular Weight:
176.30
UNSPSC Code:
12352200

form

liquid

reaction suitability

reaction type: C-H Activation
reagent type: catalyst

Application

1-Cyclohexyl-2-norbornene is a norbornene ligand developed in the lab of Guanbing Dong for us in norbornene/palladium catalysis. The catalytic system of 1-Cyclohexyl-2-norbornene, [PdCl(allyl)]2 (683396), and RuPhos (663131) can be used for the site-selective ortho functionalization of aryl iodides.1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service