Skip to Content
Merck
All Photos(1)

Documents

900377

Sigma-Aldrich

(±)-α-Tocopherol-(trimethyl-13C3 phenyl)

≥99 atom % 13C, ≥96% (CP)

Synonym(s):

DL-all-rac-α-Tocopherol-(trimethyl-13C3 phenyl), Vitamin E (trimethyl-13C3 phenyl)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C3C26H50O2
Molecular Weight:
433.68
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

≥99 atom % 13C

Quality Level

Assay

≥96% (CP)

form

liquid

technique(s)

mass spectrometry (MS): suitable

color

yellow to brown

mass shift

M+3

storage temp.

−20°C

SMILES string

CC(C)CCCC(C)CCCC(C)CCCC1(C)CCC2=C([13CH3])C(O)=C([13CH3])C([13CH3])=C2O1

InChI

1S/C29H50O2/c1-20(2)12-9-13-21(3)14-10-15-22(4)16-11-18-29(8)19-17-26-25(7)27(30)23(5)24(6)28(26)31-29/h20-22,30H,9-19H2,1-8H3/i5+1,6+1,7+1

InChI key

GVJHHUAWPYXKBD-SVFBATFISA-N

General description

(±)-α-Tocopherol is a racemic mixture of α-tocopherol which is also known as vitamin E. It is a lipid-soluble antioxidant found in whole grains, nuts, and oils. It is known to scavenge reactive oxygen species by interacting with lipids during oxidative stress.
(±)-α-Tocopherol-(trimethyl-13C3 phenyl) is an isotope of vitamin E wherein all the three methyl carbons attached to the phenyl ring are replaced by 13C6 carbon.

Application

(±)-α-Tocopherol-(trimethyl-13C3 phenyl) can be used as a stable isotope internal standard for accurate data interpretation in specific biological samples.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

ANTIOXIDANTS-Role of Antioxidant Nutrients in Defense Systems
Encyclopedia of food sciences and nutrition, 282-289 (2003)
The role of alpha-tocopherol in plant stress tolerance
Munne-Bosch S
Journal of Plant Physiology, 162(7), 743-748 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service