Skip to Content
Merck
All Photos(1)

Documents

900335

Sigma-Aldrich

Adipic-13C6 acid

≥99 atom % 13C, ≥98% (CP)

Synonym(s):

Hexanedioic-13C6 acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6H10O4
Molecular Weight:
152.10
UNSPSC Code:
12352106
PubChem Substance ID:

isotopic purity

≥99 atom % 13C

Assay

≥98% (CP)

form

crystals

mass shift

M+6

SMILES string

O[13C]([13CH2][13CH2][13CH2][13CH2][13C](O)=O)=O

InChI

1S/C6H10O4/c7-5(8)3-1-2-4-6(9)10/h1-4H2,(H,7,8)(H,9,10)/i1+1,2+1,3+1,4+1,5+1,6+1

InChI key

WNLRTRBMVRJNCN-IDEBNGHGSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

384.8 °F - closed cup

Flash Point(C)

196 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service