Skip to Content
Merck
All Photos(1)

Key Documents

900151

Sigma-Aldrich

Molybdenum oxide nanoparticle ink

greener alternative

Synonym(s):

MoO3 Dispersion, MoO3 ink, MoO3 nanoparticle ink, MoO3 suspension

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352303
NACRES:
NA.23
Pricing and availability is not currently available.

form

dispersion

Quality Level

composition

Solid content, 2.3-2.7 wt. % (crystalline MoO3 in ethanol)

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

particle size

10-40 nm

viscosity

1-3 cP

greener alternative category

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". Click here for more information.

Application

MoO3 nanoparticle ink is for slot-dye, spin coating or doctor blading for use in organic electronics as a hole injection layer or hole transport layer.

Caution

  • Store in a dark at room temperature.
  • Prior to application: shake then ultrasonicate with sonic horn and (optionally) filter through 0.45 μm PTFE filter.
  • Post-treatment: use plasma cleaning of dry films for removal of organic dispersant.

Legal Information

Product of Avantama Ltd.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

57.2 °F

Flash Point(C)

14 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yiling Wang et al.
ACS applied materials & interfaces, 7(13), 7170-7179 (2015-03-21)
Solution-processed organic-inorganic hybrids composing of MoO3 nanoparticles and PEDOT:PSS were developed for use in inverted organic solar cells as hole transporting layer (HTL). The hybrid MoO3:PEDOT:PSS inks were prepared by simply mixing PEDOT:PSS aqueous and MoO3 ethanol suspension together. A

Articles

Recent advancements in paper-based sensing platforms offer cost-effective clinical diagnostics with microfluidic channels and colorimetric or electrochemical detection zones.

Recent advancements in paper-based sensing platforms offer cost-effective clinical diagnostics with microfluidic channels and colorimetric or electrochemical detection zones.

Recent advancements in paper-based sensing platforms offer cost-effective clinical diagnostics with microfluidic channels and colorimetric or electrochemical detection zones.

Recent advancements in paper-based sensing platforms offer cost-effective clinical diagnostics with microfluidic channels and colorimetric or electrochemical detection zones.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service