Skip to Content
Merck
All Photos(1)

Key Documents

809926

Sigma-Aldrich

1α,25-Dihydroxyvitamin D3-26,26,26,27,27,27-d6 solution

100 μg/mL in ethanol, ≥98 atom % D, ≥95% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27D6H38O3
CAS Number:
Molecular Weight:
422.67
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

≥98 atom % D

Quality Level

Assay

≥95% (CP)

form

liquid

concentration

100 μg/mL in ethanol

technique(s)

mass spectrometry (MS): suitable

color

colorless

mass shift

M+6

storage temp.

−20°C

SMILES string

C=C1[C@@H](O)C[C@H](O)C/C1=C/C=C2C(CCC3[C@H](C)CCCC(O)(C([2H])([2H])[2H])C([2H])([2H])[2H])[C@]3(C)CCC/2

InChI

1S/C27H44O3/c1-18(8-6-14-26(3,4)30)23-12-13-24-20(9-7-15-27(23,24)5)10-11-21-16-22(28)17-25(29)19(21)2/h10-11,18,22-25,28-30H,2,6-9,12-17H2,1,3-5H3/b20-10+,21-11-/t18-,22-,23?,24?,25+,27-/m1/s1/i3D3,4D3

InChI key

GMRQFYUYWCNGIN-LPTLAYIMSA-N

Related Categories

General description

1α,25-Dihydroxyvitamin D3 [1,25(OH)2D3], also known as calcitriol, plays a role in calcium and phosphate metabolism in humans. It also regulates insulin secretion and induces cellular differentiation.
1α,25-Dihydroxyvitamin D3-26,26,26,27,27,27-d6 is a deuterated vitamin D3 wherein C-26 and C-27 carbon methyl protons are replaced by deuterium.

Application

1α,25-Dihydroxyvitamin D3-26,26,26,27,27,27-d6 can be used as an analytical standard for the quantitative analysis of dihydroxyvitamin D3 concentration in human serum via LC-MS.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F

Flash Point(C)

14 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantification of 1, 25-dihydroxyvitamin D2 and D3 in serum using liquid chromatography-tandem mass spectrometry
Clinical Applications of Mass Spectrometry in Biomolecular Analysis, 63(2), 291-300 (2016)
The emerging role of vitamin D3 in women′s health
Khadilkar, SS
The Journal of Obstetrics and Gynecology of India, 63(2), 147?150-147?150 (2013)
Paola Illesca et al.
Food & function, 11(10), 9086-9102 (2020-10-08)
Nutritional interventions are promising tools for the prevention of obesity. The n-3 long-chain polyunsaturated fatty acid (n-3 LCPUFA) docosahexaenoic acid (DHA) modulates immune and metabolic responses while the antioxidant hydroxytyrosol (HT) prevents oxidative stress (OS) in white adipose tissue (WAT).
Vitamin D3: a helpful immuno-modulator
Di R, et al.
Immunology, 134(2), 123-139 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service